Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007114.2 | NP_009045.2 | 16, 17 | 3388 | ||
XM_011534056.2 | XP_011532358.1 | 16, 17 | 3461 | |||
GenBank mRNA | AB209109.1 | - | 16, 17 | 3358 | ||
AK299020.1 | - | 17, 18 | 3048 | |||
AK312697.1 | - | 16, 17 | 3342 | |||
BC117418.1 | - | 16, 17 | 3241 | 40125969 | ||
BC126123.1 | - | 16, 17 | 3241 | 8991839 | ||
BC143840.1 | - | 16, 17 | 3250 | 9052355 | ||
L01042.1 | - | 16, 17 | 3141 |