Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201270.1 | NP_958427.1 | 4 | 397 | ||
GenBank mRNA | AY496861.1 | - | 3 | 307 | ||
X16058.2 | - | 4 | 397 |