Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166116.1 | NP_001159588.1 | 8 | 911 | ||
NM_003275.3 | NP_003266.1 | 8 | 988 | |||
GenBank mRNA | AK095748.1 | - | 5 | 655 | ||
AK314533.1 | - | 8 | 911 | |||
BC002660.2 | - | 8 | 988 | 3610014 | ||
M77016.1 | - | 7 | 809 |