Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000016.1 | NP_001000016.1 | 302 |
Gene Symbol: LOC100911479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750834.3 | XP_003750882.1 | 303 |
Gene Symbol: LOC103690280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767750.1 | XP_008765972.1 | 1 | 303 |