Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032283.2 | NP_001027454.1 | 3 | 716 | ||
NM_001032284.2 | NP_001027455.1 | 3 | 716 | |||
NM_001307975.1 | NP_001294904.1 | 3 | 716 | |||
NM_003276.2 | NP_003267.1 | 3 | 716 | |||
XM_005269132.3 | XP_005269189.1 | 3 | 707 | |||
XM_017019914.1 | XP_016875403.1 | 3 | 408 | |||
GenBank mRNA | AB209297.1 | - | 3 | 653 | ||
AF070631.1 | - | 3 | 657 | |||
AK315221.1 | - | 3 | 646 | |||
BC053675.1 | - | 3 | 662 | 6170639 | ||
EF028063.1 | - | 3 | 437 | |||
S76736.1 | - | 3 | 621 | |||
U09086.1 | - | 3 | 625 | |||
U09087.1 | - | 3 | 658 | |||
U09088.1 | - | 3 | 658 |