Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034124.1 | NP_001029296.1 | 6 | 588 | ||
XM_006246476.2 | XP_006246538.1 | 5 | 739 | |||
GenBank mRNA | BC100616.1 | - | 6 | 588 | 7933452 |
Gene Symbol: LOC102553715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246560.2 | XP_006246622.1 | 739 | |||
XM_006246561.2 | XP_006246623.1 | 666 |