Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271597.1 | NP_001258526.1 | 4 | 619 | ||
NM_001271598.1 | NP_001258527.1 | 4 | 619 | |||
GenBank mRNA | FM120380.1 | - | 4 | 619 | ||
FN805189.1 | - | 4 | 574 | |||
FQ214628.1 | - | 4 | 601 | |||
FQ214964.1 | - | 4 | 599 | |||
FQ215164.1 | - | 4 | 600 | |||
FQ215595.1 | - | 4 | 600 | |||
FQ216481.1 | - | 4 | 601 | |||
FQ216493.1 | - | 4 | 600 | |||
FQ216936.1 | - | 4 | 600 | |||
FQ224852.1 | - | 4 | 600 | |||
FQ224951.1 | - | 4 | 599 |