Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039377.1 | NP_001034466.1 | 13 | 1833 | ||
GenBank mRNA | AY903231.1 | - | 13 | 1833 | ||
AY903232.1 | - | 13 | 1833 | |||
AY903233.1 | - | 13 | 1833 |