Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308394.1 | NP_001295323.1 | 3 | 257 | ||
NM_003278.2 | NP_003269.2 | 3 | 354 | |||
XM_017007116.1 | XP_016862605.1 | 2 | 315 | |||
XM_017007117.1 | XP_016862606.1 | 2 | 880 | |||
GenBank mRNA | AK312093.1 | - | 3 | 362 | ||
BC011024.1 | - | 3 | 319 | 4042921 | ||
BI826174.1 | - | 3 | 257 | |||
BI826652.1 | - | 3 | 223 | |||
BI837909.1 | - | 3 | 438 | |||
CD616729.1 | - | 3 | 328 | |||
CR541981.1 | - | 3 | 258 | |||
CR542009.1 | - | 3 | 258 | |||
CR749292.1 | - | 3 | 4236 | |||
X64559.1 | - | 3 | 351 |