Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009623.1 | NP_001009623.1 | 3, 4 | 629 | ||
XM_008767797.1 | XP_008766019.1 | 3, 4 | 1367 | |||
XM_008767801.1 | XP_008766023.1 | 3, 4 | 1367 | |||
GenBank mRNA | BC087135.1 | - | 3, 4 | 629 | 7123934 | |
FQ215356.1 | - | 3, 4 | 399 |