Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005530.1 | NP_001005530.1 | 4 | 1396 | ||
GenBank mRNA | BC081902.1 | - | 4 | 1396 | 7110851 |