Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105805.1 | NP_001099275.1 | 2, 3 | 298 | ||
GenBank mRNA | BC159408.1 | - | 2, 3 | 226 | 8362552 | |
FQ211488.1 | - | 2, 3 | 269 |