Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000025.1 | NP_001000025.1 | 1 | 608 |
Gene Symbol: LOC103693363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767910.1 | XP_008766132.1 | 1 | 599 |
Gene Symbol: Olr1466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000024.1 | NP_001000024.1 | 1 | 599 | ||
XM_008767806.1 | XP_008766028.1 | 2 | 1917 |
Gene Symbol: Olr1468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000724.1 | NP_001000724.1 | 1 | 599 | ||
GenBank mRNA | AF091567.1 | - | 410 | |||
M64392.1 | - | 1 | 599 |
Gene Symbol: Olr1481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000527.1 | NP_001000527.1 | 1 | 599 | ||
GenBank mRNA | M64379.1 | - | 1 | 23 |