Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006291.2 | NP_006282.2 | 6 | 1374 | ||
XM_006720243.3 | XP_006720306.1 | 8 | 2015 | |||
XM_011537112.2 | XP_011535414.1 | 9 | 2147 | |||
XM_011537113.2 | XP_011535415.1 | 8 | 1547 | |||
XM_011537114.2 | XP_011535416.1 | 8 | 2016 | |||
XM_017021616.1 | XP_016877105.1 | 8 | 1542 | |||
GenBank mRNA | AK302929.1 | - | 4 | 322 | ||
BC128449.1 | - | 6 | 786 | 40122621 | ||
BC128450.1 | - | 4 | 572 | |||
M92357.1 | - | 6 | 1374 |