Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000034.1 | NP_001000034.1 | 1 | 281 |
Gene Symbol: Olr1512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001350.1 | NP_001001350.1 | 1 | 281 |