Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000035.1 | NP_001000035.1 | 1 | 396 |
Gene Symbol: Olr1514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000036.1 | NP_001000036.1 | 1 | 396 |