Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000038.1 | NP_001000038.1 | 563 |
Gene Symbol: LOC100909754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750855.1 | XP_003750903.1 | 1 | 563 |
Gene Symbol: Olr1515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001098.1 | NP_001001098.1 | 1 | 563 |