Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105809.2 | NP_001099279.1 | 5 | 541 | ||
GenBank mRNA | BC166808.1 | - | 5 | 541 | 7124384 | |
FQ228687.1 | - | 5 | 557 |