Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105819.1 | NP_001099289.1 | 7, 8 | 1136 | ||
XM_006246980.2 | XP_006247042.1 | 6, 7 | 1173 | |||
XM_006246981.1 | XP_006247043.1 | 7, 8 | 1862 | |||
XM_006246982.2 | XP_006247044.1 | 7, 8 | 1198 | |||
XM_008767963.1 | XP_008766185.1 | 6, 7 | 973 |