Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105819.1 | NP_001099289.1 | 6 | 1005 | ||
XM_006246980.2 | XP_006247042.1 | 5 | 1042 | |||
XM_006246981.1 | XP_006247043.1 | 6 | 1731 | |||
XM_006246982.2 | XP_006247044.1 | 6 | 1067 | |||
XM_008767963.1 | XP_008766185.1 | 5 | 842 |