Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB1 SIRNA Assays ›
Gene Symbol:
APBB1
Gene Name:
amyloid beta precursor protein binding family B member 1
Gene Aliases:
FE65, MGC:9072, RIR
Chromosome Location:
Chr.11: 6395124 - 6419830 on Build GRCh38
UniGene ID:
Hs.372840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1427
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164.4
NP_001155.1
8
1430
NM_001257319.2
NP_001244248.1
7
757
NM_001257320.2
NP_001244249.1
7
845
NM_001257321.2
NP_001244250.1
8
893
NM_001257323.2
NP_001244252.1
7
757
NM_001257325.2
NP_001244254.1
7
653
NM_001257326.2
NP_001244255.1
7
853
NM_145689.2
NP_663722.1
8
1430
NR_047512.2
8
1430
XM_006718209.3
XP_006718272.2
8
1480
XM_006718210.3
XP_006718273.2
8
1515
XM_006718211.1
XP_006718274.1
8
1686
XM_006718212.3
XP_006718275.2
8
1480
XM_006718213.1
XP_006718276.1
8
1671
XM_006718214.3
XP_006718277.2
8
1480
XM_006718215.3
XP_006718278.3
8
1788
XM_011520037.2
XP_011518339.1
7
1166
XM_011520038.2
XP_011518340.1
7
671
XM_011520040.2
XP_011518342.1
7
668
XM_011520042.1
XP_011518344.1
7
661
XM_017017639.1
XP_016873128.1
8
1686
XM_017017640.1
XP_016873129.1
8
1686
XM_017017641.1
XP_016873130.1
8
1671
XM_017017642.1
XP_016873131.1
7
1166
GenBank mRNA
AF394214.1
-
8
1389
AK293452.1
-
9
934
AK293550.1
-
7
851
AK293554.1
-
7
621
AK293613.1
-
7
485
AK293643.1
-
7
845
AK293711.1
-
6
464
AK294495.1
-
7
655
AK295241.1
-
7
722
AK295425.1
-
7
642
AK295837.1
-
7
687
AK297542.1
-
2
52
AK297544.1
-
9
957
AK297550.1
-
8
894
AK308559.1
-
6
977
AK308782.1
-
7
699
AK309000.1
-
7
758
AK309434.1
-
7
675
AK309445.1
-
7
841
AK311091.1
-
8
741
AK311101.1
-
7
856
AK311149.1
-
7
686
AK311413.1
-
7
762
AK311725.1
-
9
853
AK315965.1
-
7
844
BC010854.2
-
8
1384
3905603
BX538185.1
-
7
667
DC306210.1
-
6
464
EF103274.2
-
7
701
L77864.1
-
8
1389
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
signal transduction
axonogenesis
response to iron ion
positive regulation of neuron projection development
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone binding
tau protein binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline