Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145829.1 | NP_001139301.1 | 6 | 281 | ||
NM_001145841.1 | NP_001139313.1 | 4 | 242 | |||
NM_003282.3 | NP_003273.1 | 6 | 273 | |||
GenBank mRNA | AW984315.1 | - | 3 | 216 | ||
BC032148.2 | - | 6 | 281 | 5020339 | ||
BI833431.1 | - | 4 | 251 | |||
BP233165.1 | - | 6 | 295 | |||
BQ926240.1 | - | 6 | 282 | |||
L21715.1 | - | 5 | 246 |