Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105834.1 | NP_001099304.1 | 7, 8 | 878 | ||
GenBank mRNA | FQ216878.1 | - | 6, 7 | 670 | ||
FQ225351.1 | - | 6, 7 | 672 | |||
FQ226279.1 | - | 6, 7 | 670 | |||
FQ229232.1 | - | 6, 7 | 668 | |||
FQ232005.1 | - | 6, 7 | 671 | |||
FQ233183.1 | - | 6, 7 | 670 | |||
FQ233508.1 | - | 6, 7 | 670 | |||
FQ233844.1 | - | 6, 7 | 670 | |||
FQ234322.1 | - | 6, 7 | 670 | |||
FQ234954.1 | - | 6, 7 | 670 | |||
FQ235000.1 | - | 6, 7 | 669 |