Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003284.3 | NP_003275.1 | 1 | 141 | ||
GenBank mRNA | BC029516.1 | - | 1 | 145 | 5272281 | |
X07948.1 | - | 1 | 174 |