Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007613.1 | NP_001007614.1 | 13 | 1507 | ||
XM_008768312.1 | XP_008766534.1 | 12 | 1496 | |||
XM_008768313.1 | XP_008766535.1 | 12 | 1419 | |||
GenBank mRNA | BC079036.1 | - | 13 | 1507 | 7114313 | |
FJ168767.1 | - | 12 | 1338 | |||
FQ227563.1 | - | 1 | 42 |