Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005875.1 | NP_001005875.1 | 11 | 1245 | ||
GenBank mRNA | BC083758.1 | - | 11 | 1245 | 7192849 | |
FQ218292.1 | - | 5 | 472 |