Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC95210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005532.3 | NP_001005532.2 | 3 | 1289 | ||
XM_006247676.2 | XP_006247738.1 | 2 | 920 | |||
XM_008768322.1 | XP_008766544.1 | 2 | 920 | |||
XM_008768323.1 | XP_008766545.1 | 2 | 790 | |||
XM_008768324.1 | XP_008766546.1 | 2 | 792 | |||
XM_008768325.1 | XP_008766547.1 | 3 | 864 | |||
XM_008768326.1 | XP_008766548.1 | 3 | 920 | |||
XM_008768327.1 | XP_008766549.1 | 2 | 704 | |||
XM_008768328.1 | XP_008766550.1 | 2 | 920 | |||
XM_008768329.1 | XP_008766551.1 | 2 | 920 | |||
XM_008768330.1 | XP_008766552.1 | 2 | 2001 | |||
GenBank mRNA | BC082057.1 | - | 2 | 650 | 7132329 | |
BC088445.1 | - | 2 | 909 | 7126436 | ||
BC099187.1 | - | 3 | 1289 | 7383366 |