Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105857.2 | NP_001099327.1 | 6 | 652 | ||
XM_008768472.1 | XP_008766694.1 | 7 | 849 | |||
XM_008768473.1 | XP_008766695.1 | 7 | 783 | |||
XM_008768474.1 | XP_008766696.1 | 7 | 773 | |||
XM_008768475.1 | XP_008766697.1 | 7 | 849 |