Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199082.2 | NP_954513.2 | 4 | 685 | ||
XM_006247879.2 | XP_006247941.1 | 5 | 841 | |||
XM_006247880.2 | XP_006247942.1 | 6 | 1665 | |||
XM_006247881.2 | XP_006247943.1 | 5 | 1382 | |||
XM_006247882.2 | XP_006247944.1 | 4 | 778 | |||
XM_006247884.2 | XP_006247946.1 | 5 | 1603 |