Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1308065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077666.1 | NP_001071134.1 | 7 | 917 | ||
XM_008768856.1 | XP_008767078.1 | 7 | 994 | |||
XM_008768858.1 | XP_008767080.1 | 7 | 1006 | |||
GenBank mRNA | BC127537.1 | - | 7 | 917 | 7135417 |