Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000042.1 | NP_001000042.1 | 1 | 404 |
Gene Symbol: LOC100910999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751087.2 | XP_003751135.1 | 1 | 404 |
Gene Symbol: Olr1567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000043.1 | NP_001000043.1 | 404 |