Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000044.1 | NP_001000044.1 | 1 | 789 |
Gene Symbol: Olr1565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000908.1 | NP_001000908.1 | 1 | 792 |