Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100651.1 | NP_001094121.1 | 3 | 411 | ||
GenBank mRNA | AB177393.1 | - | 3 | 411 | ||
DY311520.1 | - | 3 | 409 | |||
FQ229771.1 | - | 444 |