Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105878.1 | NP_001099348.1 | 23 | 6681 | ||
XM_006248390.2 | XP_006248452.1 | 20 | 6681 | |||
XM_006248391.2 | XP_006248453.1 | 20 | 7090 |