Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047846.1 | NP_001041311.1 | 6 | 518 | ||
XM_006248241.2 | XP_006248303.1 | 5 | 491 | |||
XM_006248242.1 | XP_006248304.1 | 5 | 489 | |||
XM_008768630.1 | XP_008766852.1 | 5 | 550 | |||
XM_008768631.1 | XP_008766853.1 | 5 | 547 | |||
GenBank mRNA | AY325209.1 | - | 6 | 518 |