Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013866.1 | NP_001013888.1 | 9, 10 | 869 | ||
GenBank mRNA | BC089826.1 | - | 9, 10 | 869 | 7321402 | |
FQ214930.1 | - | 9, 10 | 875 | |||
FQ219915.1 | - | 9, 10 | 870 |