Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000049.1 | NP_001000049.1 | 1 | 667 |
Gene Symbol: LOC100909884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751033.1 | XP_003751081.1 | 673 |
Gene Symbol: Olr1558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000725.1 | NP_001000725.1 | 1 | 667 |
Gene Symbol: Olr1560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001009.1 | NP_001001009.1 | 1 | 667 |