Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243891.1 | NP_001230820.1 | 5 | 451 | ||
NM_001243892.1 | NP_001230821.1 | 4 | 397 | |||
NM_001243895.1 | NP_001230824.1 | 5 | 526 | |||
NM_003288.3 | NP_003279.2 | 5 | 526 | |||
NM_199359.2 | NP_955391.1 | 4 | 466 | |||
NM_199360.2 | NP_955392.1 | 5 | 526 | |||
NM_199361.2 | NP_955393.1 | 4 | 466 | |||
NM_199362.2 | NP_955394.1 | 5 | 526 | |||
NM_199363.2 | NP_955395.1 | 4 | 466 | |||
XM_011529034.2 | XP_011527336.1 | 5 | 526 | |||
XM_011529035.2 | XP_011527337.1 | 5 | 526 | |||
XM_017028038.1 | XP_016883527.1 | 4 | 451 | |||
GenBank mRNA | AF004430.1 | - | 5 | 464 | ||
AK055068.1 | - | 4 | 425 | |||
AK223031.1 | - | 5 | 438 | |||
AK293349.1 | - | 5 | 348 | |||
AK297064.1 | - | 4 | 424 | |||
AK298366.1 | - | 4 | 382 | |||
AK314501.1 | - | 5 | 484 | |||
AU124850.1 | - | 5 | 455 | |||
BC006804.1 | - | 5 | 411 | 3446037 | ||
BI667351.1 | - | 4 | 278 | |||
BI754201.1 | - | 4 | 353 | |||
BM473873.1 | - | 5 | 483 | |||
BP347669.1 | - | 4 | 364 | |||
BT019631.1 | - | 5 | 388 | |||
CR542036.1 | - | 5 | 388 | |||
CR749208.1 | - | 4 | 365 | |||
DA547106.1 | - | 5 | 427 | |||
DA871584.1 | - | 5 | 526 | |||
DC362382.1 | - | 4 | 382 | |||
DC428780.1 | - | 5 | 381 |