Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099758.1 | NP_001093228.1 | 4 | 340 | ||
XM_006248042.2 | XP_006248104.1 | 3 | 411 | |||
XM_006248043.2 | XP_006248105.1 | 4 | 430 | |||
GenBank mRNA | BC166404.1 | - | 4 | 430 | 9099247 | |
CF978233.1 | - | 4 | 305 |