Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000038.5 | NP_000029.2 | 16 | 4436 | ||
NM_001127510.2 | NP_001120982.1 | 17 | 4544 | |||
NM_001127511.2 | NP_001120983.2 | 14 | 4510 | |||
GenBank mRNA | AB210001.1 | - | 17 | 4508 | ||
M73548.1 | - | 15 | 4392 | |||
M74088.1 | - | 15 | 4369 |