Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105898.1 | NP_001099368.1 | 2 | 291 | ||
XM_006247994.2 | XP_006248056.1 | 2 | 332 | |||
XM_006247995.2 | XP_006248057.1 | 2 | 332 |