Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cggbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105900.1 | NP_001099370.1 | 3 | 808 | ||
XM_006247975.2 | XP_006248037.1 | 3 | 865 | |||
XM_008768528.1 | XP_008766750.1 | 2 | 1132 |