Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103693605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768923.1 | XP_008767145.1 | 2 | 1871 | ||
XM_008768925.1 | XP_008767147.1 | 2 | 1871 | |||
XM_008768926.1 | XP_008767148.1 | 2 | 1871 |
Gene Symbol: Car15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105901.1 | NP_001099371.1 | 2 | 153 | ||
XM_008768868.1 | XP_008767090.1 | 2 | 1871 | |||
XM_008768869.1 | XP_008767091.1 | 2 | 1871 |