Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099479.1 | NP_001092949.1 | 2 | 441 |
Gene Symbol: Vom2r59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099466.1 | NP_001092936.1 | 2 | 441 |
Set Membership: |
G-protein coupled receptor |