Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099466.1 | NP_001092936.1 | 4 | 1328 |
Gene Symbol: Vom2r60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099480.1 | NP_001092950.1 | 5 | 1349 |
Set Membership: |
G-protein coupled receptor |