Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000367.3 | NP_000358.1 | 2 | 226 | ||
XM_011514839.2 | XP_011513141.1 | 2 | 184 | |||
XM_017011241.1 | XP_016866730.1 | 3 | 520 | |||
GenBank mRNA | AF021876.1 | - | 1 | 49 | ||
AF021877.1 | - | 1 | 49 | |||
AK310420.1 | - | 2 | 120 | |||
BC005339.1 | - | 2 | 136 | 3829338 | ||
BC009596.1 | - | 2 | 135 | 4133029 | ||
BI598325.1 | - | 2 | 211 | |||
DB481604.1 | - | 2 | 230 | |||
S62904.1 | - | 1 | 114 | |||
U12387.1 | - | 2 | 125 |