Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013867.1 | NP_001013889.1 | 16 | 2122 | ||
XM_006248858.2 | XP_006248920.1 | 17 | 2949 | |||
XM_006248859.2 | XP_006248921.1 | 17 | 2817 | |||
XM_006248860.2 | XP_006248922.1 | 17 | 2949 | |||
XM_006248861.2 | XP_006248923.1 | 16 | 2858 | |||
GenBank mRNA | BC081935.1 | - | 16 | 2122 | 7114952 |