Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Get4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163320.1 | NP_001156792.1 | 4 | 459 | ||
XM_006248927.2 | XP_006248989.1 | 4 | 2695 |