Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003292.2 | NP_003283.2 | 34 | 5118 | ||
GenBank mRNA | U69668.1 | - | 34 | 4909 | ||
X66397.1 | - | 34 | 5118 |