Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102546368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769103.1 | XP_008767325.1 | 2 | 182 |
Gene Symbol: Vom2r62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099509.1 | NP_001092979.1 | 329 |
Gene Symbol: Vom2r63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099499.1 | NP_001092969.1 | 329 |
Set Membership: |
G-protein coupled receptor |